missing translation for 'onlineSavingsMsg'
Learn More

Staurosporine, Fisher BioReagents

£137.94 - £959.38

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym staurosporine, kinome_3629
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10042804
View Documents
Fisher BioReagents
100 μg Poly Micro Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10002854
View Documents
Fisher BioReagents
BP2541-500
500 μg Poly Micro Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10647155
View Documents
Fisher Bioreagents
BP2541-1
1 mg Poly Micro Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Staurosporine is a potent protein kinase inhibitor useful for studies on protein phosphorylation in the regulation of cellular functions.
Specifications

Chemical Identifiers

62996-74-1
466.541
staurosporine, kinome_3629
CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
C28H26N4O3
HKSZLNNOFSGOKW-ZYSRIHRCSA-N
49831000

Specifications

62996-74-1
White
C28H26N4O3
staurosporine, kinome_3629
Pass Test
466.541
466.5
Solid
270°C
> 98.0 %
15, 8928
HKSZLNNOFSGOKW-ZYSRIHRCSA-N
CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
49831000
>98.0%
Staurosporine, From Streptomyces sp.
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
Limited evidence of a carcinogenic effect. May be harmful by inhalation, in contact with skin and if swallowed. Moisture sensitive. Sensitivity to light. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Use personal protective equipment. Do not breathe dust. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:1
Flammability:1
Instability:1

Recommended Storage : 4°C, protect from light.

Product Identifier
  • Staurosporine

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2

Hazard Statement
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications