missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STCH Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Brand: Invitrogen PA19555
This item is not returnable.
View return policy
Description
PA1-9555 detects STCH in human samples. PA1-9555 has been successfully used in Western blot procedures. By Western blot, PA1-9555 detects a 55 kDa protein representing STCH. PA1-9555 immunogen is a purified recombinant STCH corresponding to the chromosome 21 gene (Human).
STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.Specifications
STCH | |
Polyclonal | |
Unconjugated | |
Hspa13 | |
1600002I10Rik; AV006182; B230217N24Rik; Heat shock 70 kDa protein 13; heat shock protein 13; heat shock protein 70 family, member 13; heat shock protein 70kDa family member 13; heat shock protein 70kDa family, member 13; heat shock protein family A (Hsp70) member 13; Hspa13; microsomal stress 70 protein ATPase core; microsomal stress-70 protein ATPase core; STCH; stress 70 protein chaperone microsome-associated 60 kDa protein; stress 70 protein chaperone microsome-associated 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kD; stress 70 protein chaperone, microsome-associated, 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kDa; stress 70 protein chaperone, microsome-associated, human homolog; stress-70 protein chaperone microsome-associated 60 kDa protein; testis secretory sperm-binding protein Li 199a | |
Chicken | |
IgY fraction | |
RUO | |
6782 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P48723 | |
Hspa13 | |
Purified recombinant STCH corresponding to the chromosome 21 gene (Human). | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgY |