Learn More
STE buffer soln., Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J60265.K3
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsSTE buffer is a DNA resuspension and storage buffer used in plasmid Kits for plasmid purification and in some plasmid supplementary protocols. Used in DNA extraction, DNA electrophoresis and many other lab protocols.
Solubility
Soluble in water.
Notes
Contains 100mM NaCl, 10mM Tris-HCl, 1mM EDTA, pH 8.0.

Specifications
STE buffer soln. | |
Liquid | |
2 L | |
Soluble in water. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.