missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Stealth RNAi™ siRNA Luciferase Reporter Control
Stealth RNAi™ siRNA Luciferase Reporter Control
Brand: Invitrogen™ 12935146
135.66 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Stealth RNAi™ siRNA Reporter Controls are ideal for use in RNA interference (RNAi) experiments to help you optimize your transfection conditions in any vertebrate cell line. These controls are: –Designed to efficiently knock-down its intended target –Not homologous to anything in vertebrate transcriptomeSpecifications
This individual control duplex is supplied as 250 μl at 20 μM and includes 1 ml of 1X RNA Annealing/Dilution Buffer. Store at -20°C. Products are guaranteed stable for 6 months when properly stored. | |
siRNA | |
Unconjugated | |
siRNA | |
Dry Ice |
Positive Control | |
RNAi | |
Stealth RNAi™ siRNA | |
250 μL | |
Luciferase |
For Research Use Only. Not for use in diagnostic procedures.