Learn More
Invitrogen™ Stealth RNAi™ siRNA Negative Control Hi GC Duplex #2
Stealth RNAi™ siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi™ siRNA duplex versus background.
Brand: Invitrogen™ 12935114
162.14 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Stealth RNAi™ siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi™ siRNA duplex versus background.
These controls are:
- Designed in 3 levels of GC content (Lo, Med, Hi) with 3 sequences available per level so researchers can match GC content to their experimental Stealth RNAi™ siRNA
- Not homologous to anything in the vertebrate transcriptome
- Tested not to induce stress response Stealth RNAi™ siRNA
Negative Control Kit contains all three controls (Lo, Med, Hi GC content-excluding sequences #2 and #3) and each is also available separately. Get the ideal negative controls for your RNAi experiment so you can assess your results accurately.
Specifications
This individual control is supplied as 250 μL at 20 μM. Store at -20°C. Products are guaranteed stable for 6 months when properly stored. |
|
siRNA | |
Unconjugated | |
siRNA | |
Dry Ice |
Negative Control | |
RNAi, Transfection | |
Stealth RNAi™ siRNA | |
250 μL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.