Learn More
Invitrogen™ Stealth RNAi™ siRNA Negative Control Lo GC
Stealth RNAi™ siRNA Negative Control Lo GC
Brand:  Invitrogen™ 12935200
147.40 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10719184
  • £220.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Stealth RNAi™ siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi™ siRNA duplex versus background. These controls are: –Designed in 3 levels of GC content (Lo, Med, Hi) with 3 sequences available per level so researchers can match GC content to their experimental Stealth RNAi™ siRNA –Not homologous to anything in vertebrate transcriptome –Tested not to induce stress response Stealth RNAi™ siRNA Negative Control Kit contains all three controls (Lo, Med, Hi GC content-excluding sequences #2 and #3) and each is also available separately. Get the ideal negative controls for your RNAi experiment so you can assess your results accurately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Supplied at 250 μl at 20 μM. Store at -20°C. Guaranteed stable for 6 months when properly stored.
siRNA
Unconjugated
siRNA
Negative Control
RNAi, Transfection
Stealth RNAi™ siRNA
250 μL