missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Stealth RNAi™ siRNA Negative Control, Med GC
Stealth RNAi™ siRNA Negative Control, Med GC
Brand: Invitrogen™ 12935300
205.82 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
Stealth™ RNAi siRNA Negative Control, Med GC, is one of three negative controls for use with Stealth™ siRNA experiments. These controls are a great way to measure the effect of your experimental Stealth™ RNAi siRNA duplex versus background. The Stealth™ RNAi siRNA Negative Control Kit contains all three controls (Lo, Med, and Hi GC content—excluding sequences #2 and #3), and each is also available separately. These controls are:About Stealth™ RNAi siRNA Negative Controls
Stealth™ RNAi Negative Control Duplexes are ideal for use in RNA interference (RNAi) experiments as a control for sequence independent effects following Stealth™ RNAi delivery in any vertebrate cell line. Each Stealth™ RNAi Negative Control Duplex is designed to minimize sequence homology to any known vertebrate transcript. The Stealth™ RNAi Negative Control Duplexes differ from one another in their GC content, and are supplied in a ready-to-use format. 1X RNA Annealing/Dilution Buffer is included for dilution of the Stealth™ RNAi stock solution, if desired.
Specifications
Contains one tube of 250 μl Stealth™ RNAi Negative Control Duplex (20 μM) and one tube of 1 ml 1X RNA Annealing/Dilution Buffer. Store at -20°C. | |
siRNA | |
Unconjugated | |
siRNA |
Negative Control | |
RNAi, Transfection | |
Stealth RNAi™ siRNA | |
250 μL |
For Research Use Only. Not for use in diagnostic procedures.