Learn More
Invitrogen™ Stealth RNAi™ siRNA Negative Control Med GC Duplex #3
Stealth RNAi™ siRNA Negative Control Med GC Duplex #3
Brand:  Invitrogen™ 12935113
Code : 50
Additional Details : Weight : 0.10500kg
Product Code. 10062863
  • £227.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
Stealth RNAi™ siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi™ siRNA duplex versus background. These controls are:
  • Designed in 3 levels of GC content (Lo, Med, Hi) with 3 sequences available per level so researchers can match GC content to their experimental Stealth RNAi™ siRNA
  • Not homologous to anything in the vertebrate transcriptome
  • Tested not to induce stress response Stealth RNAi™ siRNA
    Negative Control Kit contains all three controls (Lo, Med, Hi GC content-excluding sequences #2 and #3) and each is also available separately. Get the ideal negative controls for your RNAi experiment so you can assess your results accurately.
  • Specifications
    Specifications
    This individual control is supplied as 250 μl at 20 μM.  Store at -20°C. Products are guaranteed stable for 6 months when properly stored.
    siRNA
    Unconjugated
    siRNA
    Dry Ice
    Negative Control
    RNAi, Transfection
    Stealth RNAi™ siRNA
    250 μL