Learn More
Medline Scientific™ Steddy-BR Trinocular Stereo Microscope With Broad Base
The Steddy Stereo Microscopec provides high quality, wide field of view images, a wide zoom range and is highly robust and extremely well built, suitable for the most demanding uses
Brand: Medline Scientific™ 7600.0000M
This item is not returnable.
View return policy
Description
- Broad base
- Super widefield 10x, FN 22 eyepieces for maximum field of view
- Wide magnification zoom range of 3.35x to 180x (6.7x to 45x as standard)
- Trinocular head with photo port
- Dual LED illumination
Specifications
Two rubber eyecups, dust cover, instruction manual and blue filter | |
6.7X to 45X | |
Dual LED |
Broad base | |
Trinocular, 45° inclined, with photo port | |
10X/20 mm Super Widefield |