Learn More
R&L Enterprises™ Steel Gas Cylinder Stands
Provides sturdy, secure support for gas cylinders in the laboratory. R&L Enterprises™ Steel Gas Cylinder Stands are strongly constructed in nylon-coated steel.
£213.00 - £290.00
Specifications
Material Steel
Product Type Gas Cylinder Stand
For Use With (Equipment) Cylinder
View More Specs
Products 2
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
12975961
View Documents
R&L Enterprises™
3604
146 mm dia. cylinder N/A N/A N/A
12985961
View Documents
R&L Enterprises™
3605
178 mm dia. cylinder N/A N/A N/A
Description
Description
  • Available in two different sizes
  • Dimension given is the maximum diameter of cylinder accomodated
Specifications
Specifications
Steel
Gas Cylinder Stand
Cylinder
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
R&L Enterprises™ Steel Gas Cylinder Stands