missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
11904762
|
RETSCH
22.458.0005 |
35 I.D. x 51.5 mm O.D. | N/A | |||||||
11914762
|
RETSCH
22.458.0004 |
35 I.D. x 40 mm O.D. |
£90.10
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
11994752
|
RETSCH
22.458.0003 |
32 I.D. x 40 mm O.D. |
£90.10
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
Specifications
Steel Ring | |
Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RETSCH Steel Ring