missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Steel Safety Canister with Screw Cap

The Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.

Brand:  asecos™ 23129

 View more versions of this product

Product Code. 17856536

  • £69.00 / Each
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With screw cap (secured against loss) overpressure valve, PTFE seal, integrated flame barrier (removable) and plastic protective ring
  • Incl. multilingual safety sticker to designate the contents
Specifications

Specifications

0.3 gal.
Yellow
6.50 in.
7.13 in.
1.59 lb.
1 L
Steel
165.00 mm
181.00 mm
0.72 kg
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Sonderangebote

Sonderangebote