missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Steel shot, non-spherical, 0.5mm (0.02 in.) dia., Thermo Scientific™
£36.40 - £81.00
Specifications
Chemical Name or Material | Steel shot |
---|---|
Name Note | 0.5mm (0.02 in.) dia. |
MDL Number | MFCD00678174 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Steel shot | |
0.5mm (0.02 in.) dia. | |
MFCD00678174 |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures