missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories

Keep your reaction station running smoothly with these parts and accessories.

£16.95 - £2385.00


Products 5
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
15487854
View Documents
Cole-Parmer™
36630-86
Reducing Sleeve
£317.00
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
 
15447854
View Documents
Cole-Parmer™
36630-87
Small Volume Test Tubes (1 mL)
£486.00
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
 
15487844
View Documents
Cole-Parmer™
36630-83
Small Volume Test Tube (3 mL)
£530.00
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
 
16498945
View Documents
Cole-Parmer™
M7438
Coolant Supply Hose This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11806752
View Documents
Cole-Parmer™
36630-58
Reflux Head
£2385.00
Each
Estimated Shipment: 01-04-2025
to see stock.
Add to basket
 
Description

Description

  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.