Learn More
STEMCELL Technologies STEMdiff™ Cerebral Organoid Maturation Kit
Culture medium kit for extended maturation of human cerebral organoids
Brand: STEMCELL Technologies 08571
Additional Details : Weight : 1.00000kg
Description
- Generate self-organized, pluripotent stem cell (PSC)-derived neural organoids with a cellular composition and structural organization representative of the developing human brain
- This simple, four-stage protocol is based on the formulation published by Lancaster et al. (Lancaster MA et al. Nature, 2013 and Lancaster MA et al. Science, 2014) to more reliably generate cerebral organoids
- Beginning with an embryoid body (EB) formation step followed by expansion of neuroepithelia, organoids generated using STEMdiff Cerebral Organoid Kit feature cortical-like regions including the ventricular zone, outer subventricular zone, intermediate zone, and cortical plate
- For extended culture periods (> 40 days), the components required for maturation can be purchased as STEMdiff Cerebral Organoid Maturation Kit (Catalog #08571)
Specifications
Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pluripotent Stem Cells | |
Cell Culture, Differentiation, Organoid Culture | |
1 Kit |