Learn More
Zeiss™ Stemi 305 Stereomicroscope with Flexible Stands
Product Code. p-7235930
Click to view available options
Base Style:
Boom Stand B
Swiveling Stand U
Unit Size:
1 set
Product Code. 17352830
Brand: Zeiss™ 4919030010000
Only null left
Add to basket
Add to basket
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
  • This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
  • The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
  • Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Base Style Boom Stand B
For Use With (Application) Biological Education, Labs and Production Environments