missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™-34 SFM (1X)
GREENER_CHOICE

Product Code. 11580356
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11580356

Brand: Gibco™ 10639011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

A serum-free medium specifically formulated to support the development of human hematopoietic cells in culture.

  • A serum-free medium specifically formulated to support the development of human hematopoietic cells in culture
  • This includes HSC and progenitor cells isolated from bone marrow, neonatal cord blood, and peripheral blood
  • StemPro-34 SFM is a flexible cell culture media that can be used for a wide variety of applications
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hematopoietic Stem Cell, Hematopoietic Cell, Bone Marrow
Classification Serum-free
Form Liquid
Product Type Stem Cell Serum Free Media (SFM)
Serum Level Serum-free
With Additives Phenol Red
Without Additives No Glutamine
Product Line StemPro
Quantity 500 mL
Endotoxin Level ≤ 10 EU/mL
Species Human
Content And Storage • 500 ml StemPro™-34 Serum Free Medium (store at 2°C to 8°C in the dark)
• 13 ml StemPro™-34 Nutrient Supplement (store at -5°C to -20°C in the dark)

Store the complete medium at 2°C to 8°C in the dark.
Culture Type Stationary Suspension Culture
Show More Show Less

For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.