missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ hESC SFM

Product Code. 10438033
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10438033

Brand: Gibco™ A1000701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

StemPro hESC SFM is a serum- and feeder-free medium specially formulated to support the growth and expansion of pluripotent stem cells (hESCs and iPSCs). It has been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities while maintaining differentiation potential.

StemPro hESC SFM is a serum- and feeder-free medium specially formulated to support the growth and expansion of pluripotent stem cells (hESCs and iPSCs). It has been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities while maintaining differentiation potential.

StemPro hESC SFM has been extensively tested and is proven to maintain pluripotency in a growing list of hESC lines, including BG01, BG02, BG03, H1, H9, and BG01V. In addition, it has been used to scale up production of hESCs to over 1 x 1010 cells. StemPro hESC SFM has been shown to support hESC and iPSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of hESCs to differentiate into all three germ line lineages.

StemPro hESC SFM components

  • StemPro hESC Supplement
  • DMEM/F-12 with GlutaMAX medium
  • Bovine serum albumin 25% (BSA)

Additional components required for complete medium (need to be purchased separately)

  • FGF BASIC (full length) REC HU (Cat. no. PHG0261)
  • 2-Mercaptoethanol (Cat. no. 21985-023)

Applications

Formulated for the growth and expansion of human embryonic and human induced pluripotent stem cells.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Embryonic Stem Cells (ESCs)
Classification Serum-free
Form Liquid
Product Type Stem Cell Media
Serum Level Serum-free
With Additives GlutaMAX, Phenol Red
Manufacturing Quality cGMP for medical devices, 21 CFR Part 820 and ISO 13485
Product Line StemPro
Quantity 1 Kit
Shipping Condition Supplement - dry ice, Basal - ambient, BSA - ambient
Species Human
Content And Storage • StemPro™ hESC Supplement, store at -5 to -20°C and protect from light
• DMEM/F-12 with GlutaMAX medium, store at 2 to 8°C and protect from light
• Bovine serum albumin (BSA) 25%, store at 2 to 8°C and protect from light

Culture Type Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.