Learn More
STEMCELL Technologies StemSpan™ Leukemic Cell Culture Kit
For culture, expansion, and drug screening of chronic and acute myeloid leukemia cells
Brand:  STEMCELL Technologies 09720
Additional Details : Weight : 0.17220kg
Product Code. 17119961
  • £1971.00 / 1 set
Estimated Shipment: 19-07-2024
to see stock.
Description
Description
  • StemSpan™ Leukemic Cell Culture Kit has been developed for the in vitro culture and expansion of malignant cells and has been tested on chronic myeloid leukemia (CML) and acute myeloid leukemia (AML) samples
  • This optimized protocol allows users to expand, culture, and use malignant cells for drug screening
  • StemSpan™ Leukemic Cell Culture Kit includes the serum-free medium SFEM II (Catalog #09605), StemSpan™ CD34+ Expansion Supplement (10X; Catalog #02691), and small molecule UM729 (Catalog #72332)
Specifications
Specifications
Cancer Cells and Cell Lines, Hematopoietic Stem and Progenitor Cells, Leukemia and Lymphoma Cells
Cell Culture; Expansion; Toxicity Assay
1 Kit