Learn More
STEMCELL Technologies StemSpan™ Leukemic Cell Culture Kit
Serum-free medium for culture and expansion of hematopoietic cells
Brand: STEMCELL Technologies 9605
Code : Z1
Additional Details : Weight : 0.10000kg
Description
- StemSpan™ Leukemic Cell Culture Kit has been developed for the in vitro culture and expansion of malignant cells and has been tested on chronic myeloid leukemia (CML) and acute myeloid leukemia (AML) samples
- This optimized protocol allows users to expand, culture, and use malignant cells for drug screening
- StemSpan™ Leukemic Cell Culture Kit includes the serum-free medium SFEM II (Catalog #09605), StemSpan™ CD34+ Expansion Supplement (10X; Catalog #02691), and small molecule UM729 (Catalog #72332)
Specifications
Cancer Cells and Cell Lines, Hematopoietic Stem and Progenitor Cells, Leukemia/Lymphoma Cells | |
Cell Culture, Expansion, Toxicity Assay | |
100 mL |