Learn More
STEMCELL Technologies StemSpan™ Spinner Flask
Flask to expand hematopoietic cells
Brand:  STEMCELL Technologies 28800
Additional Details : Weight : 0.16670kg
Product Code. 17158071
  • POA / 1 set
Estimated Shipment: 09-08-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • The StemSpan™ Spinner Flask is a borosilicate glass flat-bottom flask with a unique agitator design including impeller blade and magnetic bar
  • The spinner flask has a 50 mL volume capacity, and is reusable and autoclavable
  • It is suitable for use with bone marrow, peripheral blood, cord blood and purified cell suspensions
Specifications
Specifications
Cell Culture
1 Ea.