Learn More
STEMCELL Technologies StemSpan™-XF
Xeno-free and serum-free medium for culture and expansion of human hematopoietic cells
Marke: STEMCELL Technologies 100-0073
Weitere Details : Weight : 0.50000kg
Beschreibung
- StemSpan™-XF has been developed for the in vitro culture and expansion of human hematopoietic cells, when the appropriate growth factors and supplements are added
- This allows users the flexibility to prepare medium that meets their requirements
- StemSpan™-XF contains pre-tested human-derived and recombinant human (rh) proteins
- Using appropriate StemSpan™ Expansion Supplements, StemSpan™-XF may be used to expand CD34+ cells isolated from human cord blood, mobilized peripheral blood, or bone marrow samples, or to expand and differentiate lineage-committed progenitor cells to generate populations of erythroid, myeloid, or megakaryocyte progenitor cells
Spezifikation
Hematopoietic Stem and Progenitor Cells | |
Cell Culture, Expansion | |
500 mL Bottle |