Learn More
R&D Systems™ StemXVivo EMT Inducing Media Supplement (100X)
To reliably induce epithelial to mesenchymal transition (EMT) in vitro
Brand:  R&D Systems™ CCM017
Product Code. 18441320
  • £560.00 / 1mL
Explore more special offers
Description
Description
Reliable induction of EMT is useful for understanding the biology of EMT and for identifying and evaluating novel prognostic markers and therapeutics for fibrotic diseases and metastasis. However, the ability to induce EMT in vitro typically requires TGF-beta stimulation or labor-intensive genetic modification. Moreover, although TGF-beta is necessary to induce EMT, it is not sufficient in all cell types.
  • Drives EMT in 5 days to ensure efficient use of time and reagents
  • Provides a straightforward method to induce EMT in vitro
  • Is completely defined to reduce experimental variability
  • May induce EMT in cells resistant to TGF-beta stimulation alone
  • Avoids time-consuming genetic modifications
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Stem Cell Supplement Media
1 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ StemXVivo EMT Inducing Media Supplement (100X) > 1mL