Learn More
R&D Systems™ StemXVivo Human/Mouse Chondrogenic Supplement, 0.5 mL
Media supplement to induce the differentiation of MSCs into chondrocytes
Brand:  R&D Systems™ CCM006
Product Code. 18401570
  • £197.00 / 0.50mL
Explore more special offers
Description
Description
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into chondrocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
  • Contains high quality differentiation factors to drive reproducible and efficient MSC chondrogenesis
  • Is defined to reduce unwanted experimental variability.
  • Has been developed and optimized using human and mouse MSCs
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
0.5 mL