Learn More
R&D Systems™ StemXVivo Human Osteogenic Supplement, 12.5 mL
Media supplement to induce the differentiation of human MSCs into osteocytes
Brand: R&D Systems™ CCM008
This item is not returnable.
View return policy
Description
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into osteocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
- Contains high quality differentiation factors to drive reproducible and efficient MSC osteogenesis
- Is defined to reduce unwanted experimental variability
- Has been developed and optimized using MSCs
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
12.5 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
R&D Systems™ StemXVivo Human Osteogenic Supplement, 12.5 mL > 12.5mL
Spot an opportunity for improvement?Share a Content Correction