Learn More
Cole-Parmer Stepless Ear Clamps
Shop All Cole-Parmer Products
Click to view available options
:
10 Stepless Ear Clamp 12 to 14.5mm
Description
Tongue-in-groove design e in., erts uniform pressure. Dimple provides increased clamping force. Fast and easy installation. Degreased to minimize contamination. Lightweight, narrow band design
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.