Learn More
Laboratoires Anios™ Steranios 2% Soaking Disinfectant
Laboratoires Anios™ Steranios 2% Soaking Disinfectant provides a high level of disinfection of medical devices, surgical, medical, endoscopic, and thermosensitive equipment.
Brand: Laboratoires Anios™ 382034
Additional Details : Weight : 21.33000kg
Description
- Ready-to-use solution
- Stability of soaking bath: 30 days
- Wide material compatibility
- High level disinfection of medical devices, surgical, medical, endoscopic and thermosensitive equipment.
- Buffered at pH6
Specifications
Disinfectant for Soaking Bath | |
Bacteria: EN 13727, EN 14561, Helicobacter py loriMycobacteria: EN 14348, EN 14563, Mycobacterium massilienseYeasts/Molds: EN 13624, EN 14562Viruses: EN 14476 : Poliovirus, Adenovirus, Norovirus MNV, HIV-1, HBV, BVDV (surrogate of HCV), HerpesvirusSpores: EN 13704 | |
Disinfection of medical devices, surgical, medical, endoscopic and thermosensitive equipment | |
5 L |
6 | |
2% glutaraldehyde solution, buffered at pH 6 in the presence of surface effects catalysor, coloring, excipients | |
4/Cs. |
Safety and Handling
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P403-Store in a well-ventilated place.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : glutaraldehyde