Learn More
Zeiss™ SteREO Discovery V8 with Coarse/Fine Drive
Click to view available options
For Use With (Equipment):
Epi-Illumination and VisiLED
Description
- The SteREO Discovery V8 offers crisp and brilliant images throughout the whole 8:1 manual zoom range.
- The noticeably improved three dimensional image perception helps you to prepare and manipulate your samples - especially in developmental biology and quality assurance.
- Acquire crisp and brilliant images throughout the whole 8:1 zoom range.
- The selectable click-stops for discrete magnification steps allow you to calibrate pixel sizes in the imaging software packages ZEN and AxioVision.
Specifications
Specifications
| For Use With (Equipment) | Epi-Illumination and VisiLED |
| For Use With (Application) | Epi-Illumination and VisiLED for Transillumination. |