missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Stericup™ E-GP Funnel-less Sterile Vacuum Filtration Systems
The newest addition to the Stericup™ Quick Release Family. Evolved with an eco-conscience.
£228.00 - £314.00
Specifications
Color | Clear with blue adapter |
---|---|
Disposable | Single-use |
Material | Polystyrene |
Material (Membrane) | PES Express PLUS |
Pore Size | 0.22 μm |
Product Code | Brand | Volume (Metric) | Outlet Fitting | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Outlet Fitting | Price | Quantity & Availability | ||||
16477545
|
Merck
SEGPU0538 |
500 mL | 38 mm threaded |
£228.00
Pack of 12 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
16487545
|
Merck
SEGPU0545 |
500 mL | 45 mm threaded |
£228.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
16497545
|
Merck
SEGPU1138 |
1000 mL | 38 mm threaded |
£261.00
Pack of 12 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
16407555
|
Merck
SEGPU1145 |
1000 mL | 45 mm threaded |
£314.00
Pack of 12 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
Description
Stericup E and Steritop E sterile vacuum systems are the newest addition to the Stericup Quick Release Family. The Stericup E and Steritop E family of devices are designed as a more sustainable option for sterile filtration that significantly reduces plastic and packaging. These are funnel-less filter products for use in the sterile vacuum filtration of aqueous solutions such as media and tissue culture reagents/buffers.The Stericup E and Steritop E devices are designed specifically for sterile filtration from compatible bottles (38 and 45 mm neck size) of media and tissue culture reagents/buffers. Like Stericup Quick Release filters, Stericup E devices feature a tactile-stop cap to signal secure closure, an ample textured writing surface, and filter membrane characteristics clearly labeled on the filter collar.SEGPU0538 combines a 38 mm threaded collar, which uses a 0.22 μm pore size fast flow PES Express PLUS membrane, with a 500 mL receiver flask for sterilizing tissue culture media and buffers. Not recommended for reagents prepared in other vessel type devices like flasks or anything without a threaded neck.Specifications
Clear with blue adapter | |
Polystyrene | |
0.22 μm | |
40 cm² | |
Bottle Top Filter | |
73 mm |
Single-use | |
PES Express PLUS | |
Sterile | |
Sterile filtration | |
73 mm |