missing translation for 'onlineSavingsMsg'
Learn More

Merck Stericup™ Quick Release-GP Sterile Vacuum Bottle Top Filtration Systems

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.

Brand:  Merck S2GPU11RE

 View more versions of this product

Product Code. 15979180

  • £348.00 / Pack of 12
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.

The Stericup™ vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.

Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device.

The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop™ bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
  • Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
  • Stable base and easy grip design of the Stericup™ device enable any user to comfortably perform all stages of the workflow in a laminar flow hood.
  • Recessed bottom of Stericup™ receiver flasks allows flasks to be stacked easily in refrigerated spaces.

Applications

  • Sterilization of Aqueous Solutions (150 mL to 1 L)
    • Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear with blue adapter
Membrane Filter
Polystyrene
Sterile
Bottle Top Filter
1000 mL
319 mm
73 mm
Single-use
PES
0.22 μm
Sterile filtration
1000 mL
319 mm
13.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Merck Stericup™ Quick Release-GP Sterile Vacuum Bottle Top Filtration Systems > 1000 mL, 1000 mL - receiver

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.