Learn More
Texwipe™ Sterile 70% Isopropanol
Evaporates with extremely low residue
Brand: Texwipe™ TX3270
257.93 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Includes: Certificates of Processing, Analysis, and Compliance
Description
Sterile solution contains 70% by volume USP grade isopropyl alcohol with 30% USP purified water. Solution is 0.2μm filtered, filled into cleaned containers, double bagged and packaged in sealed polybag case liner, and gamma-irradiated to a sterility level of 10-6.
- Evaporates with extremely low residues
- Eliminates mixing, filtration, sterilization, processing and QC expense of in-house production of sterile alcohol/water blends
- Each container is lot coded with an expiration date
Specifications
Liquid | |
Clear | |
70% USP Isopropyl Alcohol and 30% USP Purified Water | |
Trigger Spray Bottle | |
Surface cleaning, wide down for pass through to controlled environments, eliminates mixing, filtration, sterilization, processing and QC expense of in-house production of sterile Alcohol/Water blends | |
16 oz. |
Sterile | |
USP | |
70% | |
0.2 μm | |
Certificates of processing, analysis and compliance |
Safety and Handling
- texwipe 70% isopropyl alcohol, sterile
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Flammable solid Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropyl alcohol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.