missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ Sterile 70% Isopropanol

Product Code. p-2912509 Shop All Texwipe Products
Click to view available options
Volume (English):
16 oz.
1 gal.
Container Type:
Trigger Spray Bottle
Unit Size:
Pack of 12
Pack of 4
This item is not returnable. View return policy

Product Code. 15752406

Brand: Texwipe™ TX3270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Certificates of Processing, Analysis, and Compliance

Evaporates with extremely low residue

Sterile solution contains 70% by volume USP grade isopropyl alcohol with 30% USP purified water. Solution is 0.2μm filtered, filled into cleaned containers, double bagged and packaged in sealed polybag case liner, and gamma-irradiated to a sterility level of 10-6.

  • Evaporates with extremely low residues
  • Eliminates mixing, filtration, sterilization, processing and QC expense of in-house production of sterile alcohol/water blends
  • Each container is lot coded with an expiration date

Specifications

Form Liquid
Sterility Sterile
Appearance Clear
Certifications/Compliance USP
Composition 70% USP Isopropyl Alcohol and 30% USP Purified Water
Concentration 70%
Container Type Trigger Spray Bottle
Filtered 0.2 μm
For Use With (Application) Surface cleaning, wide down for pass through to controlled environments, eliminates mixing, filtration, sterilization, processing and QC expense of in-house production of sterile Alcohol/Water blends
Includes Certificates of processing, analysis and compliance
Volume (English) 16 oz.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • texwipe 70% isopropyl alcohol, sterile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Flammable solid Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : isopropyl alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.