missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Sterile CyChlor
BIOPHARMA

Provided ready-to-use and is efficacious against bacteria and yeasts in 3 mins. Contec™ Sterile CyChlor Solution is a new broad spectrum disinfectant designed for every day use.

£217.00 - £241.00

Specifications

Product Type Disinfectant
Form Liquid
pH 3 to 6
Sterility Sterile
Appearance Colorless
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15959875
BIOPHARMA
View Documents Promotion Details
Contec™
SBC100CC
1 L
£217.00
6 bottles
Please to purchase this item. Need a web account? Register with us today!
15969875
BIOPHARMA
View Documents Promotion Details
Contec™
SBC500CC
5 L
£241.00
2 bottles
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Broad spectrum efficacy in 3 mins saves time spent on biodecontamination
  • Filtered to 0.2 microns and filled in a Grade A environment ensuring the product is free from contamination and particulates
  • Suitable for Grade A and B cleanrooms
Specifications

Specifications

Disinfectant
3 to 6
Colorless
Grade A and B cleanrooms
Fluid
Limited-use
No
Liquid
Sterile
Not Autoclavable
Clear
Capped Container
Filtered
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Contec™ Sterile CyChlor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.