Learn More
Spectrum Labs™ Sterile Hollow Fiber Filter Modules
Brand: Spectrum Labs™ C02-E070-05-S
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
Hollow Fiber | |
Sterile | |
For Use with 1XFL mPES 70 Applications | |
Filter Modules |
70 kDa | |
20 cm2 | |
1 Each | |
0.5 mm |