Learn More
Pro-Lab Diagnostics™ Sterile Inoculation Loop
Product Code. 11989044
Click to view available options
Diameter (Metric) Inside:
3.57 mm
5.05 mm
Unit Size:
Pack of 25
Product Code. 11989044
Brand: Pro-Lab Diagnostics™ PL.100
Only null left
Add to basket
Add to basket
For inoculation with nichrome wire loops. Prolab Diagnostics™ Sterile Inoculation Loops are part of a complete range of inoculation tools including traditional calibrated nichrome wire loops with a selection of insulated holders.
  • Accurately calibrated nichrome wire loops
  • Standard gauge
  • Supplied in packs of 25
Specifications
Volume 10 μL
Diameter (Metric) Inside 5.05 mm
Material Nichrome
Sterility Sterile
Product Type Inoculating Loop
For Use With (Equipment) Laboratory and Research use
Quantity 25 Pk.
Calibrated Yes
For Use With (Application) Laboratory and Research use