missing translation for 'onlineSavingsMsg'
Learn More

Technical Service Consultants™ Sterile Inoculation Loop

Product Code. p-8000472
Click to view available options
Volume:
1 μL
5 μL
10 μL
Color:
Clear
Green
Red
White
Yellow
Quantity:
1000 Pk.
20/bag
50 bags of 20
50/bag
Unit Size:
Pack of 1000

Product Code. 12371309

Brand: Technical Service Consultants™ TS/2A

Only null left
Add to basket
Edge
Add to basket

Use the Technical Service Consultants™ Sterile Inoculation Loop for general purpose use or routine samples. Its soft HI polystyrene composition prevents the head breaking off, which makes it ideal for clinical use.

  • Laboratory use only
  • Shelf Life: 3 years.
  • Sterilization Method: Gamma Irradiation to achieve a SAL of 10-6
  • 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container

Specifications

Disposable Single-use
Volume 10 μL
Material Polystyrene
Sterility Sterile
Color Green
Product Type Inoculating Loop
For Use With (Equipment) Laboratory use only
Quantity 20/bag
For Use With (Application) Laboratory use only
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.