missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Service Consultants™ Sterile Inoculation Needle
Use disposable Technical Service Consultants™ Sterile Inoculation Needle to save time compared to heat sterilization of re-usable needles. The needle is perfect for counting or isolating pathogens from heavy mixed cultures.
£247.00 - £550.00
Specifications
Material | Polystyrene |
---|---|
Sterility | Sterile |
Disposable | Single-use |
Product Type | Inoculating Loop |
For Use With (Equipment) | General laboratory use only |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12391309
|
Technical Service Consultants™
TS/3-A |
Clear |
£247.00
Pack of 1000 |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
12719885
|
Technical Service Consultants™
TS/3-C |
White |
£550.00
Pack of 1000 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
Description
- Tapered tip for improved bead security
- Safe and secure transfer of cultures from freezer to culture plate
- Eliminates need to balance beads with inoculating loop
Specifications
Polystyrene | |
Single-use | |
General laboratory use only | |
General Laboratory use only |
Sterile | |
Inoculating Loop | |
1000/Pk. |