missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Sterile Plastic Inoculation Loops (Volume: 1μL)

Product Code. p-7112469
missing translation for 'orderingAttributeHoverText'
Quantity:
50 Pouches of 20
800 Pouches of 20
Packaging Type:
Microseal Pouch
Microseal Pouch, Ecopack
Microseal bags
Sterility:
Aseptically Manufactured
Gamma-irradiated
missing translation for 'unitSize'
Pack of 1000
Pack of 16000
This item is not returnable. View return policy

Product Code. 15762115

missing translation for 'mfr': Microspec™ C1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation.

  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Medium rigidity
  • Culture loops (Length: 200mm)
  • Smooth edges to avoid tearing and digging in agar
  • Volume: 1μL
  • Color: Blue

Specifications

Disposable Single-use
Volume 1 μL
Diameter (English) Inside 0.04 in.
Diameter (Metric) Inside 1 mm
Length (English) 7.87 in.
Length (Metric) 200 mm
Material High-impact Polystyrene
Sterility Aseptically Manufactured
Color Blue
Product Type Inoculating Product
Length (English) Handle 7.87 in.
Length (Metric) Handle 200 mm
Quantity 800 Pouches of 20
Packaging Type Microseal Pouch, Ecopack
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.