missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Sterile Plastic Inoculation Loops (Volume: 10μL)
Retrieves an inoculum from culture of microorganisms.
Brand: Microspec™
Additional Details : Weight : 18.95000kg
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Medium rigidity
- Culture loops (Length: 200mm)
- Smooth edges to avoid tearing and digging in agar
- Volume: 10μL
- Color: Blue
Specifications
No | |
Nickel Chromium Wire | |
Non-sterile | |
90 to 100 mm | |
Blue | |
Wire Loop Handle | |
7.87 in. | |
16000 | |
Yes |
4.15 mm | |
3.54 to 3.94 in. | |
0.163 in. | |
10 μL | |
Plastic Culture Loops | |
25 wire loops in a vial | |
200 mm | |
Plastic Vial |