missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Sterile Plastic Inoculation Loops (Volume: 10μL)

Retrieves an inoculum from culture of microorganisms.

Brand:  Microspec™

Additional Details : Weight : 17.36100kg

 View more versions of this product

Product Code. 15782105

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Medium rigidity
  • Culture loops (Length: 200mm)
  • Smooth edges to avoid tearing and digging in agar
  • Volume: 10μL
  • Color: Blue
Specifications

Specifications

Yes
High-impact Polystyrene
Sterile
200 mm
Blue
7.87 in.
800 Pouches of 20
No
4.5 mm
7.87 in.
0.18 in.
10 μL
Plastic Culture Loops
200 mm
Microseal Pouch
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers