Learn More
Microspec™ Sterile Plastic Inoculation Loops (Volume: 5μL)
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture
Brand: Microspec™ B4G
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Medium rigidity
- Culture loops (Length: 200mm)
- Smooth edges to avoid tearing and digging in agar
- Volume: 5μL
- Color: Blue
Specifications
Single-use | |
2.5 mm | |
200 mm | |
Gamma-irradiated | |
Plastic Culture Loops | |
200 mm | |
Microseal Pouch |
0.10 in. | |
7.87 in. | |
High-impact Polystyrene | |
Blue | |
7.87 in. | |
50 Pouches of 20 | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.