missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Sterile Plastic Inoculation Loops (Volume: 5μL)
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture
£48.85 - £546.00
Specifications
Diameter (English) Inside | 0.10 in. |
---|---|
Diameter (Metric) Inside | 2.5 mm |
Length (English) | 7.87 in. |
Length (Metric) | 200 mm |
Material | High-impact Polystyrene |
Product Code | Brand | Quantity | Packaging Type | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Sterility | Price | Quantity & Availability | |||||
15712115
|
Microspec™
B1 |
50 Pouches of 20 | Grip Seal Bag | Aseptically Manufactured | N/A | ||||||
15752115
|
Microspec™
B4G |
50 Pouches of 20 | Microseal Pouch | Gamma-irradiated |
£58.65
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||||
15742115
|
Microspec™
B4 |
50 Pouches of 20 | Microseal Pouch | Aseptically Manufactured |
£48.85
Pack of 1000 |
Estimated Shipment: 14-04-2025
Log in to see stock. ![]() |
|||||
15722115
|
Microspec™
B1G |
800 Pouches of 20 | Microseal Pouch | Sterile | N/A | ||||||
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Medium rigidity
- Culture loops (Length: 200mm)
- Smooth edges to avoid tearing and digging in agar
- Volume: 5μL
- Color: Blue
Specifications
0.10 in. | |
7.87 in. | |
High-impact Polystyrene | |
Blue | |
200 mm |
2.5 mm | |
200 mm | |
Single-use | |
7.87 in. | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Microspec™ Sterile Plastic Inoculation Loops (Volume: 5μL)