Learn More
Corning™ Sterile Polypropylene Straight Containers with Polyethylene Caps
Click to view available options
60 mL; Sterile; White Cap
Description
- Injection molded polypropylene
- Leakproof polyethylene screw cap
- Assured Sterile 10-3
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity (Metric) | 60 mL |
Color | Clear |
Material | Polypropylene |
No. per Case | 700/Cs. |
No. per Pack | 700/Pk. |
Sterility | Sterile |
Closure Color | White |