Learn More
Corning™ Sterile Polystyrene Inoculating Loops
GREENER_CHOICE
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture. Gosselin™ Sterile Polystyrene Inoculating Loops provide a superior solution for transferring microscopic organisms, inoculum, or streaking.
Brand:  Corning™ OS10B-02
 View more versions of this product
Product Code. 11769515
  • £444.09 / Pack of 9000
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polystyrene (PS) inoculating loops
  • Spherical or needle extremity
  • 1 μl or 10 μl
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 μL
Polystyrene
Blue
150 BG
195 mm
Sterile
Inoculating Loop