Learn More
Corning™ Sterile Polystyrene Inoculating Loops
GREENER_CHOICE
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture. Gosselin™ Sterile Polystyrene Inoculating Loops provide a superior solution for transferring microscopic organisms, inoculum, or streaking.
Brand:  Corning™ OS10B-02
 View more versions of this product
Product Code. 12810165
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Polystyrene (PS) inoculating loops
  • Spherical or needle extremity
  • 1 μl or 10 μl
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 μL
Polystyrene
Blue
3000/Pk.
195 mm
Sterile
Inoculating Loop, Spherical Tip
Tamper evident zip bag