Learn More
Fisherbrand™ Sterile Rayon Tip Transport Swabs
For the collection and/or transportation of a wide range of bacteria, viruses and facultative microorganisms.
Brand: Fisherbrand™ 300285
This item is not returnable.
View return policy
Description
- All transport swabs have an inert, non-toxic rayon tip
- Gamma radiation sterilized
- Lot number and expiration date printed on labels
Specifications
ISO 9002/EN 29002 | |
3 shelves | |
Sterile | |
Swab |
Round | |
100Pack | |
Plastic | |
Rayon |