Learn More
SP Bel-Art™ Sterileware Cross-Sectional Liquid Samplers
The integrated valve of this sterile tool closes as it is pushed against a container bottom securing the sample for withdrawal. Sample can then be poured from the top of the tool into your sample container.
Brand:  SP Bel-Art™ F37874-1100
 View more versions of this product
Product Code. 16816926
  • £277.20 / Pack of 20
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Provides effective cross-sectional sampling of liquids and light creams
  • Reaches bottom of container while keeping hand clear of container top
  • Diameter allows for use through drum bunghole or other small openings
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) samplers heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
225 mL
20/Pk.
2.1 cm
110 cm