Learn More
Bel-Art™ Sterileware™ Sterile Styrene Sampler Spoons
Polystyrene plastic sterile sampler spoons are designed to gather small amounts of sample material.
£55.35 - £423.00
Specifications
Product Type Spoon
Material Polystyrene
Autoclavable Not Autoclavable
Includes Certificate of sterilization
Handle Type Long
View More Specs
Products 8
Product Code Brand Capacity (Metric) Quantity Price Quantity & Availability  
Product Code Brand Capacity (Metric) Quantity Price Quantity & Availability  
11761566
View Documents
Bel-Art™
H36946-0000
14.79 mL 200/Cs.
£367.00
Pack of 200
In Stock
to see stock.
Add to basket
15500142
View Documents
Bel-Art™
H36940-0010
1.23 mL 10/Pk.
£60.95
Pack of 10
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
11378733
View Documents
Bel-Art™
H36942-0000
2.46 mL 200/Cs.
£423.00
Pack of 200
In Stock
to see stock.
Edge
Add to basket
15530142
View Documents
Bel-Art™
H36946-0010
14.79 mL 10/Pk.
£55.60
Pack of 10
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
10573924
View Documents
Bel-Art™
H36940-0000
1.23 mL 200/Cs.
£407.00
Pack of 200
In Stock
to see stock.
Add to basket
15510142
View Documents
Bel-Art™
H36942-0010
2.46 mL 10/Pk.
£60.70
Pack of 10
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
15520142
View Documents
Bel-Art™
H36944-0010
4.93 mL 10/Pk.
£55.35
Pack of 10
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11316944
View Documents
Bel-Art™
H36944-0000
4.93 mL 200/Cs.
£381.00
Pack of 200
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
Description
Description
Manufactured from polystyrene plastic. SP Scienceware™ Sterileware™ Sterile Styrene Sampler Spoons are designed to gather small amounts of sample material.
  • Sterile, individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
  • FDA grade materials; safe for food, drug and cosmetic contact
  • Long handle reaches easily into jars, bottles and other confined areas, and minimizes the potential for personal contact with sampled material
  • Overall length 17.8 cm (7 in.)
Specifications
Specifications
Spoon
Not Autoclavable
Long
Sterile
FDA
7 in.
Polystyrene
Certificate of sterilization
Single-use
White
Designed to gather small amounts of sample material
17.8 cm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Bel-Art™ Sterileware™ Sterile Styrene Sampler Spoons