missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP Bel-Art™ Sterileware Upright Handle Dippers/Ladles
Available in blue or white to allow for effective sample contrast, these sterile ladles are useful for sampling liquids and creams
Brand: SP Bel-Art™ F36775-0050
Additional Details : Weight : 16.00000kg
Description
- Upright handle allows for sampling from above container
- Curved handle end provides secure withdrawal of sample
- Fits into 2 in. openings such as a 45-gallon drum bunghole
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- White high density polyethylene (HDPE), or blue polypropylene (PP) ladles heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifica
50 mL | |
Blue | |
40/Pk. | |
26 cm |