Learn More
SP Bel-Art™ Sterileware Volumetric Sampling Spoons
Suitable for use with powders, granules, liquids, gels or suspensions, these sterile spoons allow set volumes to be accurately measured and sampled
£127.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16816916
|
SP Bel-Art™
F36943-0001 |
1 mL |
£127.00
Pack of 100 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
16826916
|
SP Bel-Art™
F36943-0005 |
5 mL |
£127.00
Pack of 100 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
16836916
|
SP Bel-Art™
F36943-0010 |
10 mL |
£127.00
Pack of 100 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
16846916
|
SP Bel-Art™
F36943-0020 |
20 mL |
£127.00
Pack of 100 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
Description
- Leveling to top of bowl provides accurately measured samples
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) spoons heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
100/Pk. |